Centring rings with filter fleece, type KF
Supplier: Welch by Gardner Denver
701213 701211 701203 701201
ILMV701213EA 35.4 EUR
ILMV701213 ILMV701211 ILMV701203 ILMV701201
Centring rings with filter fleece, type KF
Seals Sealing Rings
Centring rings made of stainless steel, with filter fleece and perbunan O-ring.
Pore width: 25 µm