Buffer, bicarbonate, 1M solution, pH 9,0

Supplier: ALFA AESAR

J60116.AK
J60116.AKEA 59.8 EUR
J60116.AK
Buffer, bicarbonate, 1M solution, pH 9,0
Buffers Biological Buffers

CAS No.: 144-55-8
Physical form: liquid
MDL No.: MFCD00003528

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR