Phosphate, 0,2 M buffer solution, pH 4,4

Supplier: ALFA AESAR

J60467.AP J60467.K2
J60467.APEA 72.9 EUR
J60467.AP J60467.K2
Phosphate, 0,2 M buffer solution, pH 4,4
Buffers Biological Buffers

CAS No.: 7558-80-7
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
MDL No.: MFCD00003527

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR