Phosphate, 0,2 M buffer solution, pH 8,0

Supplier: ALFA AESAR

J60480.AP
J60480.APEA 77.9 EUR
J60480.AP
Phosphate, 0,2 M buffer solution, pH 8,0
Buffers Biological Buffers

CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527

Order Now

SPECIFICATIONS

Environmentally Preferable
Physical form Liquid
CAS No. 7558-80-7
MDL No. MFCD00003527

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR