Tricine-buffered saline (5X), pH 8,0

Supplier: ALFA AESAR

J60745.AK J60745.AP
J60745.AKEA 70.1 EUR
J60745.AK J60745.AP
Tricine-buffered saline (5X), pH 8,0
Buffers Biological Buffers

CAS No.: 5704-04-1
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00004277

Order Now

SPECIFICATIONS

Environmentally Preferable
Physical form Liquid
CAS No. 5704-04-1
MDL No. MFCD00004277
Appearance Colourless

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR