SSPE (20X), RNAse free
Supplier: ALFA AESAR
J60783.AP
J60783.APEA 0 EUR
J60783.AP
SSPE (20X), RNAse free
Buffers Electrophoresis Buffers Hybridisation Buffers
SSPE (20X) is a standard reagent in Southern and Northern hybridisation procedures. This liquid can be used straight or diluted as needed.
Contains 0,2 M sodium phosphate, 3,0 M sodium chloride, 20 mM EDTA.