BLOTTO and Tween 20 in PBS

Supplier: ALFA AESAR

J61880.AP
J61880.APEA 0 EUR
J61880.AP
BLOTTO and Tween 20 in PBS
Buffers Electrophoresis Buffers Blocking Buffers

Storage Temperature: −30 to −10 °C
Physical Form: Liquid


Applications: Blocking reagent in immunohistochemistry experiments

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR