Buffer, tris(hydroxymethyl)aminomethane hydrochloride, 1 M solution, pH 7,4, RNase free

Supplier: ALFA AESAR

J62848.AK
J62848.AKEA 82.1 EUR
J62848.AK
Buffer, tris(hydroxymethyl)aminomethane hydrochloride, 1 M solution, pH 7,4, RNase free
Buffers Cloning Buffers

CAS No.: 1185-53-1
Physical form: liquid
Appearance: colourless
MDL No.: MFCD00012590

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR