Buffers, sodium borate, 0,5 M solutions

Supplier: ALFA AESAR

J60068.AK J60068.AP J62902.AK J62902.AP J63637.AK J63637.AP
J60068.AKEA 74.9 EUR
J60068.AK J60068.AP J62902.AK J62902.AP J63637.AK J63637.AP
Buffers, sodium borate, 0,5 M solutions
Buffers Biological Buffers

CAS No.: 1330-43-4
MDL No.: MFCD00081185

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR