IEF Anode buffer (50X)

Supplier: ALFA AESAR

J63303.AP J63303.K2
J63303.APEA 36.5 EUR
J63303.AP J63303.K2
IEF Anode buffer (50X)
Buffers Electrophoresis Buffers Running Buffers

Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD03846400
Applications: For isoelectric focusing (IEF)

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR