Software for Julabo units
Supplier: JULABO GmbH
8900005 8900110 8901105
JULA8900005EA 1340 EUR
JULA8900005 461-3618 461-0063
Software for Julabo units
Software Laboratory Software
A range of software that allows programming, viewing and recording of temperature and time dependent processes in combination with laboratory instruments. Comprehensive communication and networking possibilities via RS232/RS485, USB (with optional converter) and Profibus assist with laboratory automation.
  • Start/stop functions and set point input with graphical zoom functions
  • Indication of valid actual, heating and cooling capacity
  • Repeat memorised profiles
  • Display of up to four curves in one diagram
  • Ramp function of up to 100 steps