Ampoule breakers
Supplier: JUST PLASTICS
1033-010
JUST1033-010EA 424 EUR
JUST1033-010
Ampoule breakers
Ampoule Openers
These ampoule breakers are made of PE.
  • Heavy wall flexible safety collars break ampoules safely and retain broken top for safe disposal
  • For ampoules 5 to 10 ml capacity
  • Easy and quick to handle