Liquid nitrogen containers, type APOLLO
Supplier: KGW
2509 2508 2507 2510 2513
KGWA2509EA 9140 EUR
KGWA2509 KGWA2508 KGWA2507 KGWA2510 KGWA2513
Liquid nitrogen containers, type APOLLO
Dewars and Cryogenic Vessels
Liquid nitrogen containers are made up of vacuum insulated stainless steel.
  • Integrated safety devices at the container neck
  • Vacuum lock with safety valve
  • Easy running castors
  • Hand and protection ring
It features with low evaporation rate by multi-layer superinsulation in the vacuum space. Very durable by robust constructive design, production and choice of material. Long holding time using adsorption and getter materials.
Equipped with mini-flange-joint NW 50 KF, transfer siphon with transfer hose covered with metallic tissue, hand and protectionMore Product Information