Funnels, powder, KIMBLE®
Supplier: KIMBLEKONTES
29020-80
KIML29020-80EA 205 EUR
KIML29020-80
Funnels, powder, KIMBLE®
Funnels Powder Funnels
Borosilicate glass KIMAX®.
  • Constructed with heavy uniform wall
  • Strong fire-polished rim and stem
  • High resistance to chemical attack, mechanical and thermal shock
  • ASTM Specification E1095, Type IV requirements
Conforms to USP Type I and ASTM E438, Type I, Class A requirements.