2-(Dimethylamino)pyridine 97%

Supplier: ALFA AESAR
Danger

L02513.06 L02513.14
L02513.06EA 36.7 EUR
L02513.06 L02513.14
2-(Dimethylamino)pyridine 97%
2-(Dimethylamino)pyridine

Purity: 97% CAS No.: 5683-33-0 Molecular Formula: C7H10N2 Formula Weight: 122.17 Flash Point: 80°(176°F) Refractive Index: 1.5590 Boiling Point: 190-192° Beilstein Reference: 110353 MDL No.: MFCD00006261

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 190…192 °C
Density: 1.008
Flash Pt: 80 °C (176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006261
CAS Number: 5683-33-0
EINECS: 227-147-5
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR