6-Chloro-o-cresol ≥98%

Supplier: ALFA AESAR
Danger

L03617.06 L03617.14
L03617.06EA 80.2 EUR
L03617.06 L03617.14
6-Chloro-o-cresol ≥98%
6-Chloro-o-cresol

Purity: 98% CAS No.: 87-64-9 Molecular Formula: C7H7ClO Formula Weight: 142.59 Flash Point: 72°(161°F) Refractive Index: 1.5450 Boiling Point: 75-76°/12mm Beilstein Reference: 774623 UN No.: 2669 MDL No.: MFCD00020000

Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 75…76 °C (12 mmHg)
Melting Pt: 0
Density: 1.201
Flash Pt: 72 °C (161 °F)
Storage Temperature: Ambient
MDL Number: MFCD00020000
CAS Number: 87-64-9
EINECS: 201-760-8
UN: 2669
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR