Dihexylamine ≥98%

Supplier: ALFA AESAR
Danger

L06326.AE L06326.AP
L06326.AEEA 55.9 EUR
L06326.AE L06326.AP
Dihexylamine ≥98%
Dihexylamine

Purity: 98+% CAS No.: 143-16-8 Molecular Formula: C12H27N Formula Weight: 185.36 Flash Point: 104°(219°F) Refractive Index: 1.4335 Melting Point: 3° Boiling Point: 236° Beilstein Reference: 1738519 UN No.: 2927 MDL No.: MFCD00009521

Formula: C₁₂H₂₇N
Boiling Pt: 236 °C
Melting Pt: 3 °C
Density: 0,790
Flash Pt: 104 °C (219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009521
CAS Number: 143-16-8
EINECS: 205-588-4
UN: 2927
ADR: 6.1,I

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR