Chloroform ≥99% stabilised

Danger


Stabilization: Stabilised with ethanol 1 %

L13200.AP L13200.AU
L13200.APEA 30.1 EUR
L13200.AP L13200.AU
Chloroform ≥99% stabilised
Chloroform

Stabilized with 0.8-1% ethanol


Purity: 99% CAS No.: 67-66-3 Molecular Formula: CHCl3 Formula Weight: 119.38 Refractive Index: 1.4440 Odour: Characteristic Boiling Point: 60.5-61.5° Beilstein Reference: 1731042 Merck Reference: 14,2141 UN No.: 1888 MDL No.: MFCD00000826

Formula: CHCl₃
Boiling Pt: 60,5…61.5 °C
Melting Pt: –63 °C
Density: 1,492
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 14,2141
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9cc00da8cefd4e04c0ea1d7c5801308cd73eeef9&vid=96870c784520df552dcde9a71610c883a1c71b50&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR