Potassium (4-methylphenyl)trifluoroborate ≥98%

Supplier: ALFA AESAR
Warning

L17604.03 L17604.06
L17604.03EA 53 EUR
L17604.03 L17604.06
Potassium (4-methylphenyl)trifluoroborate ≥98%
Potassium (4-methylphenyl)trifluoroborate

Purity: 98% CAS No.: 216434-82-1 Molecular Formula: C7H7BF3K Formula Weight: 198.04 Melting Point: 300° Beilstein Reference: 8290098

Formula: C₇H₇BF₃K
MW: 198.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318171
CAS Number: 216434-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR