Bromotrispyrrolidinophosphonium hexafluorophosphate ≥97%

Supplier: ALFA AESAR
Warning

L19384.03 L19384.06
L19384.03EA 42.8 EUR
L19384.03 L19384.06
Bromotrispyrrolidinophosphonium hexafluorophosphate ≥97%
Bromo-tris-pyrrolidinophosphonium hexafluorophosphate

Purity: 97% CAS No.: 132705-51-2 Molecular Formula: C12H24BrF6N3P2 Formula Weight: 466.19 Storage Temperature: -30°C to -10°C

Formula: C₁₂H₂₄BrF₆N₃P₂
MDL Number: MFCD00077412
CAS Number: 132705-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR