Heating thermostats with transparent baths, ECO
Supplier: LAUDA
L001096 L001097 L001098 L001099 L001100 L001101 L001102 L001103 L001125 L001126 L001127 L001128 L001129 L001130 L001131 L001154 L001155 L001156 L001157 L001158 L001159 L001160 L001161
461-1288EA 3370 EUR
461-1288 461-1236 LAUDL001098 461-1239 461-1289 461-1237 461-1238 461-1240 LAUDL001125 LAUDL001126 LAUDL001127 LAUDL001128 LAUDL001129 LAUDL001130 LAUDL001131 LAUDL001154 LAUDL001155 LAUDL001156 LAUDL001157 LAUDL001158 LAUDL001159 LAUDL001160 LAUDL001161
Heating thermostats with transparent baths, ECO
Circulating Thermostats Open Circulator Systems Baths with Detachable Circulators
ECO units with transparent plastic baths provide visibility in all cases where test samples need to be observed during thermostating. The thermostats with polycarbonate baths can be used in the temperature range of up to 100 °C. They have a filling volume of 5 up to 20 litres. The 15 litre (or larger) Silver and Gold baths accommodate up to three capillary viscometers. The Silver and Gold ET 6, ET 12 and ET 20 models are equipped with a cooling coil as standard. Bath/circulation thermostats have menu navigation in plain text for user friendly operation. Various interface modules areMore Product Information