Cooling thermostats, ECO Series
Supplier: LAUDA
L001333 L001335 L001336 L001337 L001338 L001339 L001341 L001342 L001343 L001344 L001357 L001359 L001360 L001361 L001362 L001363 L001365 L001366 L001367 L001368 L001381 L001382 L001383 L001384 L001385 L001386 L001387 L001388 L001389 L001390 L001391 L001392 L001393 L001394 L001395 L001396 L001397 L001398 L001399 L001400 L001401 L001402 L001403 L001404 L001260
461-1256EA 7040 EUR
461-1256 461-1260 461-1262 461-1264 461-1266 461-1257 461-1261 461-1263 461-1265 461-1267 LAUDL001357 LAUDL001359 LAUDL001360 LAUDL001361 LAUDL001362 LAUDL001363 LAUDL001365 LAUDL001366 LAUDL001367 LAUDL001368 LAUDL001381 LAUDL001382 LAUDL001383 LAUDL001384 LAUDL001385 LAUDL001386 LAUDL001387 LAUDL001388 LAUDL001389 LAUDL001390 LAUDL001391 LAUDL001392 LAUDL001393 LAUDL001394 LAUDL001395 LAUDL001396 LAUDL001397 LAUDL001398 LAUDL001399 LAUDL001400 LAUDL001401 LAUDL001402 LAUDL001403 LAUDL001404 LAUDL001260
Cooling thermostats, ECO Series
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Cooling thermostats with menu navigation in plain text for user friendly operation. Various interface modules are available as accessories. All models are available as air or water-cooled versions. They are similar in specifications with the ECO model thermostats, in addition these models comes with natural refrigerant.
  • Variopump with six adjustable stages
  • Flow rate switch for control of flow rate between internal and external circulation
  • Simultaneous display of set and actual temperature
  • Over-temperature setting via menu
  • USB interface as standard