Nessler reagent, tablets

Supplier: TINTOMETER

511261BT 511231BT 511230BT 511251BT 511250BT
511261.EA 133 EUR
511261. LOVI511231BT LOVI511230BT LOVI511251BT LOVI511250BT
Nessler reagent, tablets
Nessler's reagent
MDL Number: MFCD00049656
CAS Number: 7783-33-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR