Urea ≥99.5%, Proteomics Grade

Supplier: VWR Chemicals

M123-1KGEA 275 EUR
M123-1KG M123-2.5KG M123-500G
Urea ≥99.5%, Proteomics Grade
Urea

Commonly used to denature nucleic acids for electrophoresis and to study the secondary and tertiary structure of proteins. Effective for maintaining DNA in a highly denatured state during sequencing protocols.

Formula: NH₂CONH₂
MW: 60.06 g/mol
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 132.7 °C
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 13,09935
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=62455afae7c29355c4ff33e9ad370faf78b9c80c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Abs.@280 nm (5 M, Water) <0.05
Biuret Test Pass
Chloride ≤0.0005%
Cyanate ≤0.002%
DNase none detected
Heavy Metals (as Pb) ≤0.0005%
Insolubles <0.005%
Iron ≤0.0001%
Melting Point 132 – 135 °C
Protease none
Purity ≥99.5%
RNase none detected
Solubility (5 M, Water) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR