Gentamicin sulphate, powder

Supplier: MEDIATECH

61-098-RA 61-098-RF
MDTC61-098-RAEA 68 EUR
MDTC61-098-RA MDTC61-098-RF
Gentamicin sulphate, powder
Gentamicin sulphate

An antibiotic agent, gentamicin inhibits protein synthesis by binding to the 30S ribosomal subunit, blocking the translocation of peptidyl-tRNA from the acceptor site to the donor site


In another mode of action, gentamicin acts on Pseudomonas aeruginosa by binding to the outer membrane of this bacterium. In doing so, it displaces natural cations, destabilizes the membrane and forms holes in the cell surface. Gentamicin acts against Gram-negative bacteria, Staphylococcus aureus and other Gram-positive bacteria.


Packaging: Shipping temperature: Ambient

Storage Temperature: Refrigerator
MDL Number: MFCD00270181
CAS Number: 1405-41-0
Merck Index: 13,04403

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR