Digital camera, Globecam
Supplier: MEDLINE SCIENTIFIC
9554.1019
MEDL9554.1019EA 830 EUR
MEDL9554.1019
Digital camera, Globecam
Cameras
This eyepiece camera can be inserted into the tube of a microscope.
  • No need of a trinocular head
  • Parfocal digital micro-image with immediate display
  • Clear image reproduction thanks to the CMOS sensor
Suitable for use with biological and stereo microscopes.