Dissolved oxygen meters, bench, SevenExcellence™ S900
Supplier: Mettler - Toledo
30092987 30092984 30092985 30092986
METT30092987EA 5480 EUR
METT30092987 METT30092984 METT30092985 METT30092986
Dissolved oxygen meters, bench, SevenExcellence™ S900
Electrochemistry Meters Oxygen Meters
SevenExcellence S900 bench top instruments provide customers with a premium bench top solution to measure dissolved oxygen (DO) in aqueous media.
  • Single-channel instrument is modularly expandable with additional measurement parameters at any time
  • Professional calibration support, incl. automatic calibration with Rondolino sample changer
  • Large colour display and intuitive menu guidance in 10 languages
  • Flexible method concept for high reproducibility and security
  • Extra security thanks to sophisticated user management and Intelligent Sensor Management (ISM®)
More Product Information