Masterflex® L/S® Standard Digital Drives, Avantor®
Supplier: Avantor Fluid Handling
MFLX07522-20-EUEA 3670 EUR
MFLX07522-20-EU MFLX07522-30-EU MFLX07522-20-UK MFLX07522-30-UK
Masterflex® L/S® Standard Digital Drives, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Designed with maintenance-free high-accuracy motor and a graphical interface—makes setup and operation easier than ever!
  • Flow rate 0.001/0.006 to 566/3400 ml/min with L/S tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating modes—continuous run, timed dispense, copy dispense, or volume dispense
  • Select your preferred language from the user-friendly menu for programming and operation
  • Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles;
More Product Information