CPC® Quick-Disconnect Fittings with Automatic Shutoff Valve, Polysulfone
Supplier: Avantor Fluid Handling
MFLX31313-01EA 298 EUR
MFLX31313-01 MFLX31313-03 MFLX31313-21 MFLX31313-22 MFLX31313-23 MFLX31313-01BLK MFLX31313-03BLK MFLX31313-21BLK MFLX31313-22BLK MFLX31313-23BLK MFLX31313-02BLK
CPC® Quick-Disconnect Fittings with Automatic Shutoff Valve, Polysulfone
Fittings and Connectors
Eliminate the need for pinch clamps.
  • Body and seal materials are made of USP Class VI-compliant and BSE/TSE-free materials
  • Valves restrict flow when fitting halves are disconnected
  • Fitting body constructed of durable polysulfone
These CPC® quick-disconnect fittings feature an automatic shutoff valve which closes off the flow path at disconnection, protecting valuable media while eliminating the need for pinch clamps. The thumb latch enables easy one-handed operation. An audible click is heard when the body and insert are coupled to confirm a secure,More Product Information