Masterflex® Correlated and Direct-Read Variable Area Flowtubes for Air and Water, Avantor®
Supplier: Avantor Fluid Handling
03217-07 03217-09 03219-50 03219-51 03219-52 03219-54 03219-55 03219-56 03219-59 03219-60 03219-66 03219-74 32047-00 32047-01 32047-02 32047-10 32047-11 32047-12 32047-13 32047-14 32047-15 32047-16 32047-18 32047-19 32047-20 32047-29 32047-32 32047-57 32047-62 32047-63 32047-64 32047-68 32047-69 32047-71 32047-72 32047-74 32047-77 32047-84 32047-86
MFLX03217-05EA 91.5 EUR
MFLX03217-05 MFLX03217-11 MFLX03217-13 MFLX03217-15 MFLX03217-17 MFLX03217-19 MFLX03217-21 MFLX03217-23 MFLX03217-29 MFLX03217-33 MFLX03219-53 MFLX03219-57 MFLX03219-58 MFLX03219-72 MFLX32047-09 MFLX32047-17 MFLX32047-21 MFLX32047-22 MFLX32047-23 MFLX32047-27 MFLX32047-28 MFLX32047-31 MFLX32047-50 MFLX32047-51 MFLX32047-60 MFLX32047-61 MFLX32047-66 MFLX32047-67 MFLX32047-70 MFLX32047-73 MFLX32047-76 MFLX32047-81 MFLX32047-88 MFLX32047-89 MFLX32047-90 MFLX03217-07 MFLX03217-09 MFLX03219-50 MFLX03219-51 MFLX03219-52 MFLX03219-54 MFLX03219-55 MFLX03219-56 MFLX03219-59 MFLX03219-60 MFLX03219-66 MFLX03219-74 MFLX32047-00 MFLX32047-01 MFLX32047-02 MFLX32047-10 MFLX32047-11 MFLX32047-12 MFLX32047-13 MFLX32047-14 MFLX32047-15 MFLX32047-16 MFLX32047-18 MFLX32047-19 MFLX32047-20 MFLX32047-29 MFLX32047-32 MFLX32047-57 MFLX32047-62 MFLX32047-63 MFLX32047-64 MFLX32047-68 MFLX32047-69 MFLX32047-71 MFLX32047-72 MFLX32047-74 MFLX32047-77 MFLX32047-84 MFLX32047-86
Masterflex® Correlated and Direct-Read Variable Area Flowtubes for Air and Water, Avantor®
Flow Meters
Create your own flowmeter systems for complete flexibility.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Use with air or water for laboratory and industrial applications
  • Required system components for flowmeter systems would be frame, flowtube(s), valve cartridge, and tripod—order each component separately
  • Minimum flow rate is
More Product Information