Masterflex® Drum Pump Air Powered Motors, Avantor®
Supplier: Avantor Fluid Handling
70604-25
MFLX70604-25EA 1120 EUR
MFLX70604-25
Masterflex® Drum Pump Air Powered Motors, Avantor®
Pumps Drum/Transfer Pumps
Safety transfer a variety of liquids from drums, tanks, and small storage vessels.
  • Maintenance is reduced with no mechanical seals
  • Run dry capability
Combine the dump pump air motors with a flowmeter (sold separately) to display current and total volume dispensed, Each motor includes a muffler. A filter, regulator, and lubricator are recommended.
Flow rate is measured under a 1" restriction.