Polypropylene (PP) High-Head Chemical-Resistance Centrifugal Pumps
Supplier: Avantor Fluid Handling
72012-00 72012-05 72012-10 72012-15 72012-20 72012-25
MFLX72012-05EA 465 EUR
MFLX72012-00 MFLX72012-05 MFLX72012-10 MFLX72012-15 MFLX72012-20 MFLX72012-25
Polypropylene (PP) High-Head Chemical-Resistance Centrifugal Pumps
Pumps Centrifugal Pumps
Twin bearing system handles thrust loads for extended service life.
  • Quiet, energy efficient, and compact
  • Internal cooling circuit reduces bearing temperature
  • Enclosed motor for use in harsh environments
  • All wetted parts are nonmetallic for excellent chemical resistance
High-head polypropylene pumps have a closed impeller design to significantly increase the output pressure. Polypropylene construction provides excellent chemical resistance in applications, such as filtration, where a high-head is needed.
Delivery information: Includes a 6-ft (1.8More Product Information