High-Precision High-Pressure Piston Pump Accessories
Supplier: Avantor Fluid Handling
41010-76 73150-91 74931-94 74931-95 74931-98
MFLX41010-76EA 288 EUR
MFLX41010-76 MFLX73150-91 MFLX74931-94 MFLX74931-95 MFLX74931-98
High-Precision High-Pressure Piston Pump Accessories
Pumps HPLC/GPC Pumps
Designed for optimal operation in your Ismatec pumps.
  • Choose from a wide range of replacement parts and accessories
The overall length of Solva 95605-series stop tubing is 16" (406 mm). The legacy Ismatec stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.