Seepex Variable-Speed Progressing Cavity Pump Heads and Pumps
Supplier: Avantor Fluid Handling
76804-13 76804-31 76804-40 76804-41 76804-43
MFLX76804-13EA 2830 EUR
MFLX76804-13 MFLX76804-31 MFLX76804-40 MFLX76804-41 MFLX76804-43
Seepex Variable-Speed Progressing Cavity Pump Heads and Pumps
Pumps Progressive Cavity Pumps
Excellent chemical resistance, pulseless flow, and low shear.
  • Gentle low-shear fluid transfer capable of pumping small particles without damaging product
  • Pumps can handle a variety of polymers and chemicals—ideal for pharmaceutical and water treatment applications
  • Sanitary models approved for 3A sanitary applications and can be cleaned in place
A
Pump heads can be mounted on a NEMA 56C frame motor. Pump heads can also be directly connected or belt driven by an AC or DC motor, gasoline engine (min. 2 hp), or air motor.
B
Pumps are close-coupled to an ACMore Product Information