Masterflex® L/S® PTFE-Diaphragm Pump System, Avantor®
Supplier: Avantor Fluid Handling
MFLX77915-10-EUEA 3640 EUR
MFLX77915-10-EU MFLX77915-10-UK
Masterflex® L/S® PTFE-Diaphragm Pump System, Avantor®
Pumps Diaphragm Pumps
Ideal for chemical feed/metering and applications requiring high-purity, high-pressure or both.
  • Flow range 80 to 800 ml/min—flow rate depends on drive rpm and tubing size
  • PTFE wetted parts help maintain fluid purity
  • Pump through filters or into pressurized system
  • Check valve on inlet side of pump holds prime for up to two hours when pump is off
  • Unique geometry minimizes fluid churning and dead volume for high metering accuracy without reagent contamination
  • Accurate and repeatable settings with 3-digit LED display of rpm
  • Maximize processes in a minimal
More Product Information