Masterflex® MasterSense™ Gear Pump Systems
Supplier: Avantor Fluid Handling
MFLX77976-50-EUEA 8620 EUR
MFLX77976-50-EU MFLX77976-52-EU MFLX77976-51-EU MFLX77976-50-UK MFLX77976-52-UK MFLX77976-51-UK
Masterflex® MasterSense™ Gear Pump Systems
Pumps Gear Pumps
Versatile gear pump systems for high-accuracy fluid delivery - with an intuitive touch-screen interface.
  • Powerful, space-efficient drives accept all Micropump A and B mount gear pump heads
  • Pulseless gear flow technology allows for high repeatability over long duration
  • High viscosity capability (0,2 to 1500 centipoise)
  • 12,7 cm (5") capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Single-point pressure monitoring provides real-time pressure display and alarms for process reliability
  • Continuous run, volume dispense, and time
More Product Information