Masterflex® Ismatec® IP and IP-N Digital Multichannel Peristaltic Pumps, Avantor®
Supplier: Avantor Fluid Handling
78010-03 78010-23 78010-33 78010-53 78011-11 78011-13 78011-23 78011-33 78011-43
MFLX78011-49-EUEA 7450 EUR
MFLX78011-49-EU MFLX78011-49-UK MFLX78010-03 MFLX78010-23 MFLX78010-33 MFLX78010-53 MFLX78011-11 MFLX78011-13 MFLX78011-23 MFLX78011-33 MFLX78011-43 MFLX78010-11 MFLX78010-13
Masterflex® Ismatec® IP and IP-N Digital Multichannel Peristaltic Pumps, Avantor®
Pumps Peristaltic Pumps
Smooth, precise flow and fewer tubing changes.
  • Planetary drive with 8 stainless steel rotors reduces pulsation, increases flow rate accuracy, and lengthens tubing life
  • Four-digit LED display facilitates repeatability
  • Control pump via 6-button keypad or analog and TTL external signals
  • Automatic occlusion cartridges are easy to use and provide reproducible results
These Ismatec® IP and IP-N digital peristaltic multichannel pumps feature planetary gears that drive eight stainless steel rollers in unison for smooth rotation and accurate liquid flow. TheMore Product Information