Masterflex® L/S® Single-Use Gamma-Irradiated Pump Tubing, Avantor®
Supplier: Avantor Fluid Handling
MFLX96119-16EA 199 EUR
MFLX96119-16 MFLX96123-15 MFLX96123-16 MFLX96119-15 MFLX96119-24 MFLX96123-24 MFLX96122-15
Masterflex® L/S® Single-Use Gamma-Irradiated Pump Tubing, Avantor®
Tubing Peristaltic Pump Tubing
Reduce the risk of microbial contamination.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Gamma sensitive labeling confirms treatment
Masterflex® gamma-More Product Information