Microplate readers, multi-mode, FilterMax™ F3 and F5
Supplier: Molecular Devices
F5 F3
MLDVF5EA 0 EUR
MLDVF5 MLDVF3
Microplate readers, multi-mode, FilterMax™ F3 and F5
Microplate Readers
The FilterMax™ series of multi-mode microplate readers can be operated as standalone instruments or can be integrated seamlessly with laboratory automation platforms. Both units are ideal for a broad range of applications, including drug discovery, genomics, proteomics and cell-based research. The patented design ensures precise performance and sensitivity across all detection modes. Units can use either automated or manual Z-Height Optimisation, and can utlise high-density well scanning for unique applications.
  • Performance driven specifications for sensitivity, dynamic range, accuracy,
More Product Information
Order Now
SPECIFICATIONS
Model FilterMax™ F5 FilterMax™ F3
Environmentally Preferable
Light source High Power LEDs and Deuterium Lamp High power LEDs
Measuring modes Endpoint, kinetic, linear scan and area scan Endpoint, kinetic, linear scan and area scan
Detector Silicon Photodiode and PMT Silicon Photodiode and PMT
Wavelength range 230 - 750 nm 340 - 650 nm
Absorbance range 0 – 3,5 OD 0 – 3,5 OD
Filters 2 EX + 1 EM filter slides (6 filter capacity) 1 EX + 1 EM filter slides (6 filter capacity)
Measurement time Reads a 96-well plate in 18 s, a 384-well plate in 37 s Reads a 96-well plate in 18 s, a 384-well plate in 37 s
Plate types 6- to 1536-well plates 96- to 384-well plates
Shaking Linear and orbital (3 speeds) Linear and orbital (3 speeds)
Software SoftMax Pro software SoftMax Pro software
Temperature range Ambient +4 to 45 °C
W×D×H 390×580×240 mm 390×580×240 mm
Weight 25 kg 25 kg