Universal anti-slip tapes
Supplier: 3M
AR1SZ25 AR1SZ50 AR1SZ100 AR1GS50 AR1SZ150 AR1GB50
MMMAAR1SZ25EA 819 EUR
MMMAAR1SZ25 MMMAAR1SZ50 MMMAAR1SZ100 MMMAAR1GS50 MMMAAR1SZ150 MMMAAR1GB50
Universal anti-slip tapes
Tapes
3M™ Universal Anti-Slip Tape is a simple and cost effective way to prevent slips and trips in the workplace.
  • Slip-resistant tape for most shoe traffic applications
  • Resistant to abrasion
  • Proven durability for long life
  • Self adhesive backing for quick and easy application
Effective adherence to clean, dry surfaces without wrinkling, curling, tearing, shrinking, or lifting. This tape can be used on a wide variety of surfaces including concrete, resilient tile, metal, and porous surfaces (if primed). It is also suitable for use on ramps, heavy-dutyMore Product Information