Respirator filters, TR-300+ Series, for Versaflo™ powered air respirator
Supplier: 3M
TR-3822E TR-3802E TR3712E 7100266123
MMMATR-3822EEA 482 EUR
MMMATR-3822E 111-2403 111-2409 MMMA7100266123
Respirator filters, TR-300+ Series, for Versaflo™ powered air respirator
Respirators Reusable Respirators Respirator Filters
These filters provide protection against dusts, mists and other particulate hazards when used with the 3M™ Versaflo Powered Air Respirator, TR-300 Series, and a 3M™ approved headtop.
  • Filters can be exchanged quickly and easily
  • Filters need replacing at required filter interval decided by risk assesment
TR-3712E - Particulate (P) filter for protection against dusts, mists, fumes and other particulate hazards.
TR-3802E - Particulate (P) filter for protection against dusts, mists, fumes and other particulate hazards + nuisance level Organic Vapours (More Product Information