Micro haematocrit capillaries, VITREX®
Supplier: Vitrex Medical A/S
167313 160813 161313 164213 161513 161613 163213 155113 164113 160513 163613 160413 162713 160213 161813LRIS 162313 161113 160613
MODU167313EA 91.8 EUR
MODU167313 720-2289 720-2680 MODU164213 720-2290 720-2291 MODU163213 MODU155113 MODU164113 MODU160513 MODU163613 720-2679 MODU162713 720-2689 MODU161813LRIS MODU162313 MODU161113 MODU160613
Micro haematocrit capillaries, VITREX®
Pipettes Microcapillaries
These tubes are supplied fire polished at the colour-coded end, which should be sealed with the Sigillum Wax. Fire polishing reduces wear on the centrifuge seals.
  • Each heparinised tube contains at least 4 IU heparin as prescribed by the Standard
  • Tubes are available in two types of packaging: Dispenser vials of 100 tubes each for hygienic and easy handling; and economy cardboard boxes, which reduce transport and handling costs
  • Sigillum Wax plate (needs to be ordered separately) has 2×24 numbered positions per pair, which makes it easy to seal and transport
More Product Information