2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid

Supplier: MOLEKULA

58714707-100G 58714707-500G
MOLE58714707-100GEA 81 EUR
MOLE58714707-100G MOLE58714707-500G
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid
2-[Bis(2-hydroxyethyl)amino]ethanesulphonic acid
Formula: C₆H₁₅NO₅S
Boiling Pt: 405 °C (1013 hPa)
Melting Pt: 152…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00007533
CAS Number: 10191-18-1
EINECS: 233-465-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR