Indol-3-acetic acid

Supplier: MOLEKULA

M12865413
MOLEM12865413EA 0 EUR
MOLEM12865413
Indol-3-acetic acid
Indol-3-acetic acid
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Boiling Pt: 120…125 °C (0.05 mmHg)
Melting Pt: 167…170 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005636
CAS Number: 87-51-4
EINECS: 201-748-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR