(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)

Supplier: MOLEKULA

M79047769
MOLEM79047769EA 0 EUR
MOLEM79047769
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
Formula: C₁₈H₂₈F₆N₆OP₂
MW: 520.4 g/mol
MDL Number: MFCD00077411
CAS Number: 128625-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR