Combined safety storage cabinets, type 90, K-PHOENIX-90 / K-PHOENIX Vol.2-90
Supplier: asecos
30333001 30334001 30335001 30336001
135-1763EA 4720 EUR
135-1763 135-1764 MSGA30335001 135-2584
Combined safety storage cabinets, type 90, K-PHOENIX-90 / K-PHOENIX Vol.2-90
Furniture Storage Cabinets
Fireproof safety cabinets, classified type 90 (fire resistant up to 90 minutes), safe, durable, robust and convenient.
Two in one: storage of flammable liquids and aggressive chemicals in one cabinet.
  • Safety elements assembled outside the storage compartment for increased protection against corrosion, easy to clean
  • Standard door open arrest system (compartment with folding doors), doors stop in any position (compartment with wing doors) - convenient and safe removal or storage of containers
  • Doors lockable with cylinder locking (integration in a locking
More Product Information