Plastic Box with White Lid
Supplier: MULTIROIR
192-3000
MULC192-3000EA 254 EUR
MULC192-3000
Plastic Box with White Lid
Boxes Storage Boxes
These combi box containers are versatile, providing ideal storage solutions for a diverse range of laboratory applications.
  • Polypropylene material
  • Watertight lid
  • Tamper-proof lid
  • Stackable box
  • Food contact quality
Ideal for laboratory environments, this versatile container ensures secure storage and transport of samples and reagents. The airtight, tamper-evident lid provides enhanced safety, while the stackable design maximises space efficiency. The opaque white polypropylene construction ensures durability and easy identification of contents.