Sterilised thio bags, Whirl-Pak®
Supplier: NASCO FORT ATKINSON
B01040WA B01254WA B01402WA B01403WA B01603WA B01601WA
NASCB01040WAEA 99.2 EUR
NASCB01040WA NASCB01254WA NASCB01402WA NASCB01403WA NASCB01603WA NASCB01601WA
Sterilised thio bags, Whirl-Pak®
Bags Sample Bags
EPA approved for more sustainable, efficient microbiological water sampling container. A cost effective, low environmental impact alternative to rigid material containers.
  • Plus 1 sterilisation, post-manufacturing sterilisation: Increased sample integrity, perforated closure to maintain sterility
  • Thiosulfate tablet in the bag, saving technician's time
  • Gusseted bottom option, for self-standing ease
  • Liquid Lock closure for secure samples every time
  • Puncture-proof closure tabs: Inner and outer tapes for longevity and stability, wire for easy opening, pouring,
More Product Information