Nucleosil 300 C4, HPLC Columns
Supplier: Hichrom Limited
NC300-5C4-10CEA 0 EUR
NC300-5C4-10C HICHNC300-5C4250AS
Nucleosil 300 C4, HPLC Columns
Chromatography Columns
These NUCLEOSIL® HPLC columns are packed with spherical porous silica available in a pore size of 300 Å. This silica is used as the base material for reversed phase bioanalytical columns.
  • Wide range of bonded phases
  • Good mechanical stability
  • Highly efficient
Surface area: 100 m²/g