Baths, stirred, digital, Clifton, NE4-D series
Supplier: NICKEL ELECTRO
NE4-8D NE4-14D NE4-22D NE4-28D NE4-38D
NE4-8DEA 2420 EUR
NE4-8D NE4-14D NE4-22D NE4-28D NE4-38D
Baths, stirred, digital, Clifton, NE4-D series
Baths Water Baths, Stirred
General purpose stirred water baths offering good temperature stability and uniformity for consistent, repeatable results. Operation below ambient is possible using an optional refrigerated dip cooler.
  • Choice of five tank sizes and models with control from –20 to +99,9 °C with optional immersion cooler
  • PID temperature control with smart technology for adaptive or optimised temperature modes
  • Continuous or timer mode from 0 to 999 min with automatic switch-off and audible alarm
  • Low liquid level protection and auto setting temperature alarms
  • Anti-bacterial
More Product Information